Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WIBG Antibody (OTI5E2), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274868V
Description
WIBG Monoclonal specifically detects WIBG in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
WIBG | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PYM1 | |
Full length recombinant protein of human WIBG (NP_115721) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI5E2 | |
Western Blot, Immunohistochemistry | |
MGC13064, partner of Y14 and mago, PYMProtein wibg homolog, within bgcn homolog (Drosophila) | |
Mouse | |
Affinity Purified | |
DNA replication Transcription Translation and Splicing | |
84305.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction