Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WIBG Monoclonal Antibody (OTI1E10)
GREENER_CHOICE

Catalog No. PIMA526798
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526798 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526798 Supplier Invitrogen™ Supplier No. MA526798
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

WIBG Monoclonal Antibody for Western Blot, IHC (P)

Key regulator of the exon junction complex (EJC), a multiprotein complex that associates immediately upstream of the exon-exon junction on mRNAs and serves as a positional landmark for the intron exon structure of genes and directs post-transcriptional processes in the cytoplasm such as mRNA export, nonsense-mediated mRNA decay (NMD) or translation. Acts as an EJC disassembly factor, allowing translation-dependent EJC removal and recycling by disrupting mature EJC from spliced mRNAs. Its association with the 40S ribosomal subunit probably prevents a translation-independent disassembly of the EJC from spliced mRNAs, by restricting its activity to mRNAs that have been translated. Interferes with NMD and enhances translation of spliced mRNAs, probably by antagonizing EJC functions. May bind RNA; the relevance of RNA-binding remains unclear in vivo, RNA-binding was detected by PubMed:14968132, while PubMed:19410547 did not detect RNA-binding activity independently of the EJC. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen WIBG
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1E10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PYM1
Gene Accession No. Q9BRP8
Gene Alias A030010B05Rik; Partner of Y14 and mago; protein wibg homolog; PYM; PYM homolog 1 exon junction complex-associated factor; PYM homolog 1, exon junction complex associated factor; Pym1; RGD1306096; WIBG; within bgcn homolog; within bgcn homolog (Drosophila)
Gene Symbols PYM1
Host Species Mouse
Immunogen Full length human recombinant protein of WIBG produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84305
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.