Learn More
DWK Life Sciences Wheaton™ Wide Mouth Bottle, Safety Coated
SDP
Catalog No. p-7111700 Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
Closure Liner:
Polyethylene
Polyvinyl
Wide mouth safety coated clear glass bottles with attached screw caps.
  • Clear, Type III soda-lime glass
  • Largest bottles available
  • Ideal for general, small to large volume storage
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 250 mL
Coating Safety Coated
Color Clear
Material Borosilicate Glass (Type 3)
Quantity 12/Case
Closure Color White
Product Type Wide Mouth Bottle
Closure Liner Polyethylene
Closure Material Polypropylene
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.