Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ UniStore™ Reagent Bottles, Wide Mouth, PP
SDPSCI_ED

Catalog No. S00629 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1000 mL
2000 mL
4000 mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S00629 4000 mL
S43629 30 mL
S43631 60 mL
S43632 125 mL
S43635 250 mL
S43636 500 mL
S43637 1000 mL
S00628 2000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. S00629 Supplier United Scientific™ Supplier No. 33314
Add to Cart
Add to Cart
This item is not returnable. View return policy

Wide Mouth Polypropylene Reagent Bottles are rigid, translucent and have fine contact clarity.

UniStore™ Wide Mouth, PP, reagent bottles are manufactured from USP Class VI resin that is FDA-21CFR compliant.

  • Excellent chemical resistance to most acids, bases, and alcohols.
  • Polypropylene caps are included.
  • Manufactured from USP Class VI resin that is FDA-21CFR compliant for food contact and free of heavy metals.
  • Leakproof closure with integrated seal ring.
  • Precision molded for consistent wall thickness with smooth interior and exterior surfaces.
  • Certificates of Analysis for leakproof test details are available upon request.
  • The 4000 mL bottle is leakproof at ambient temperature and pressure.

Specifications

Capacity 4000 mL (135.3 oz.)
Capacity (Metric) 4000 mL
Capacity (English) 135.3 oz.
Color Translucent White
For Use With (Application) Liquids and Solutions
Material Polypropylene
Quantity 1 Ea.
Sterility Non-sterile
Width (Metric) 228.6 mm
Product Line UniStore
Closure Included Yes
Graduated No
Mouth Wide
Height (Metric) 279 mm
Product Type Reagent Bottle
Height (English) 11 in.
Closure Material Polypropylene
Diameter (Metric) 22.9 cm
Through the Closure Dispensing No
Autoclavable Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.