Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WIF-1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | WIF-1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP1794920
![]() |
Novus Biologicals
NBP17949120UL |
20 μL |
Each for $206.00
|
|
|||||
NBP179491
![]() |
Novus Biologicals
NBP179491 |
100 μL |
Each for $487.50
|
|
|||||
Description
WIF-1 Polyclonal specifically detects WIF-1 in Human samples. It is validated for Western Blot.Specifications
WIF-1 | |
Polyclonal | |
Rabbit | |
Signal Transduction, Wnt Signaling Pathway | |
WIF-1, WNT inhibitory factor 1 | |
WIF1 | |
IgG | |
38 kDa |
Western Blot | |
Unconjugated | |
RUO | |
NP_009122 | |
11197 | |
Synthetic peptide directed towards the N terminal of human WIF1The immunogen for this antibody is WIF1. Peptide sequence RKAQQRMPAIPVNIHSMNFTWQAAGQAEYFYEFLSLRSLDKGIMADPTVN. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title