Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WIF-1 Antibody (RM0144-3M51), Allophycocyanin, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP122520APC
Description
WIF-1 Monoclonal antibody specifically detects WIF-1 in Mouse samples. It is validated for Western BlotSpecifications
WIF-1 | |
Monoclonal | |
APC | |
PBS | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot | |
RM0144-3M51 | |
Western Blot | |
WIF-1, WNT inhibitory factor 1 | |
Mouse recombinant protein of WIF-1 | |
0.1 mL | |
Signal Transduction, Wnt Signaling Pathway | |
11197 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction