Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WIF-1 Antibody (RM0144-3M51) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP1225200.025MG
Description
WIF-1 Monoclonal specifically detects WIF-1 in Mouse samples. It is validated for Western Blot.Specifications
WIF-1 | |
Monoclonal | |
LYOPH | |
Western Blot 1:500-1:1000 | |
WIF-1, WNT inhibitory factor 1 | |
Rat | |
Protein G purified | |
RUO | |
11197 | |
Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG2 |
Western Blot | |
RM0144-3M51 | |
Unconjugated | |
A 0.2 um filtered solution in PBS with No Preservative | |
WIF1 | |
Mouse recombinant protein of WIF-1 | |
0.025 mg | |
Primary | |
This was produced from a hybridoma (mouse myeloma) fused with spleen cells from a rat immunized with mouse recombinant protein of WIF-1. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction