Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    WIF-1 Antibody (RM0144-3M51), mFluor Violet 500 SE, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP122520MFV500
Description
WIF-1 Monoclonal antibody specifically detects WIF-1 in Mouse samples. It is validated for Western BlotSpecifications
| WIF-1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified | 
| Western Blot | |
| RM0144-3M51 | |
| Western Blot | |
| WIF-1, WNT inhibitory factor 1 | |
| Mouse recombinant protein of WIF-1 | |
| 0.1 mL | |
| Signal Transduction, Wnt Signaling Pathway | |
| 11197 | |
| Store at 4C in the dark. | |
| IgG2 | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            