Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WIF-1 Rabbit anti-Human, Mouse, Rat, Clone: 1X6C9, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP316580100UL
This item is not returnable.
View return policy
Description
WIF-1 Monoclonal antibody specifically detects WIF-1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
WIF-1 | |
Monoclonal | |
Unconjugated | |
WIF-1, WNT inhibitory factor 1 | |
A synthetic peptide corresponding to a sequence within amino acids 280-379 of human WIF-1 (Q9Y5W5). | |
100 μg | |
Signal Transduction, Wnt Signaling Pathway | |
11197 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
1X6C9 | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction