Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WIF1 Monoclonal Antibody (1G5)
GREENER_CHOICE

Catalog No. PIMA515701
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515701 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515701 Supplier Invitrogen™ Supplier No. MA515701
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15701 targets WIF1 in indirect ELISA, IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-15701 immunogen is purified recombinant fragment of human WIF1 expressed in E. Coli. MA5-15701 detects WIF1 which has a predicted molecular weight of approximately 42kDa.

The protein encoded by this gene functions to inhibit WNT proteins, which are extracellular signaling molecules that play a role in embryonic development. This protein contains a WNT inhibitory factor (WIF) domain and five epidermal growth factor (EGF)-like domains, and is thought to be involved in mesoderm segmentation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WIF1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1G5
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene WIF1
Gene Accession No. Q9WUA1, Q9Y5W5
Gene Alias AW107799; UNQ191/PRO217; WIF; WIF1; WIF-1; Wnt inhibitory factor 1
Gene Symbols WIF1
Host Species Mouse
Immunogen Purified recombinant fragment of human WIF1 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11197, 24117
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.