Learn More
WIF1 Polyclonal Antibody, Invitrogen™
Description
This antibody is predicted to react with mouse, rat, Xenopus and zebrafish based on sequence homology.
Specifications
Specifications
| Antigen | WIF1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide |
| Gene | WIF1 |
| Gene Accession No. | Q9Y5W5 |
| Gene Alias | AW107799; UNQ191/PRO217; WIF; WIF1; WIF-1; Wnt inhibitory factor 1 |
| Gene Symbols | WIF1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.