Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wijs solution, Volumetric, 0.1 M ICl, Honeywell Fluka™
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations

Specifications
Specifications
Boiling Point | 118°C |
CAS Min % | ≥90.0000% |
CAS Max % | ≤100.0000%, <2.0000% |
Quantity | 6 x 1 L |
Linear Formula | ICl |
MDL Number | MFCD00011354 |
UN Number | UN2789 |
Beilstein | 3587194 |
Flash Point | 40°C |
Synonym | Iodine monochloride solution, Iodine according to Wijs for the iodine value determination, Chloroiodide solution, Wijs solution |
Show More |
Safety and Handling
ShelfLife | 450 days from date of manufacture |
DOTInformation | Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Acetic acid solution |
Recommended Storage | Room Temp |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction