Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ WIKI4
GREENER_CHOICE

Catalog No. 485525
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-552-5 25 mg
48-555 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 48-552-5 Supplier Tocris Bioscience™ Supplier No. 4855/25
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent tankyrase inhibitor

WIKI4 is an inhibitor of Wnt/β-catenin signaling (EC50 ∼ 75 nM). Inhibits auto-ADP-ribosylation of tankyrase 2 (TNKS2) (IC50 ∼15 nM); prevents the ubiquitination and degradation of axin. WIKI4 inhibits expression of β-catenin target genes in cancer cell lines as well as in human embryonic stem cells (ESC).

WIKI4 synthesized to Ancillary Material Grade also available.

Specifications

Chemical Name or Material 2-[3-[[4-(4-Methoxyphenyl)-5-(4-pyridinyl)-4H-1,2,4-triazol-3-yl]thio]propyl]-1H-benz[de]isoquinoline-1,3(2H)-dione
CAS 838818-26-1
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C29H23N5O3S
Formula Weight Observed MW: 521.59
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.