Description
- Will not melt, drip, or ignite at 350° for five minutes
- Adjustable side vent to ensure increased airflow
- Fits naturally to face contours to block out airborne contaminants
- Lightweight and comfortable with a pivot headband that allows optimal placement
- Antifog lenses
Compliance
Meet ANSI™ Z87.1-2010 standards.
Specifications
Specifications
| Lens Coating | Anti-fog |
| Certifications/Compliance | ANSI Z87.1-2010 |
| Lens Tint | Clear |
| Product Type | Heat-resistant Safety Goggles |
| Size | Universal |
| Ventilation Details | Adjustable side vent |
| UV Protection | UV400 |