Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ William's E Medium, GlutaMAX™ Supplement
GREENER_CHOICE

Catalog No. p-7076330
Encompass_Preferred
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format.

William's E Medium was originally developed by Williams and Gunn as reduced serum-supplemented medium for long-term cell cultures of adult rat liver epithelial cells. Gibco™ William's E Medium can also be used for growing hepatocyte cells of different species, such as human derived HepaRG™ cells.

This William's E Medium is modified as follows:

  • With: Phenol Red, GlutaMAX™
  • Without: HEPES

The complete formulation is available.

Gibco™ William's E Medium is a modification of earlier media that has been enriched in amino acids and double the glucose. William's E Medium contains unique ingredients, including zinc, iron, manganese, non-essential amino acids, the reducing agent glutathione and the lipid methyl linoleate. Gibco™ William's E Medium with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format.

cGMP Manufacturing

Gibco™ William's E Medium is manufactured at a cGMP compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

William's E Medium contains no proteins or growth factors. Therefore, William's E Medium requires supplementation, commonly with 5-10% Fetal Bovine Serum (FBS). William's E Medium uses a sodium bicarbonate buffer system (2.2 g / L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Mammalian Cells
Form Liquid
Product Type William's E Medium
With Additives GlutaMAX
Without Additives No HEPES
Product Line GlutaMAX™
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2°C to 8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Culture Type Mammalian Cell Culture
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.