Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wilms Tumor Protein Polyclonal Antibody, Biotin, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS6983RBIOTIN

This item is not returnable.
View return policy
Description
Wilms Tumor Protein Polyclonal antibody specifically detects Wilms Tumor Protein in Bovine, Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western BlotSpecifications
Wilms Tumor Protein | |
Polyclonal | |
Biotin | |
WT1 | |
AWT1; D630046I19Rik; EWS-WT1; GUD; NPHS4; WAGR; Wilms tumor 1; Wilms tumor 1 homolog; Wilms tumor protein; Wilms tumor protein homolog; Wilms tumor protein isoform Ex4a(+); WIT1; WIT-1; WIT-2; WT; Wt1; Wt-1; WT1 antisense RNA; WT1AS; WT1-AS; WT1-AS1; WT33 | |
KLH conjugated synthetic peptide derived from human Wilms Tumor Protein | |
100 μL | |
Primary | |
Bovine, Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300 | |
P19544, P22561, P49952 | |
Rabbit | |
Protein A | |
RUO | |
22431, 24883, 7490, 782183 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction