Learn More
Description
Specifications
Specifications
| Antigen | Wilms Tumor (WT33) |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | Please refer to lot specific datasheet. |
| Conjugate | Unconjugated |
| Gene Symbols | WT1 |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated linear peptide corresponding to 16 amino acids from the N-terminal region of human Wilms Tumor Protein. The immunogen is conserved in isoforms 1,2,3 and 4. |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.