Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ WIN 18446
GREENER_CHOICESDP

Catalog No. 473650 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-365-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 47-365-0 Supplier R&D Systems™ Supplier No. 4736/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. ALDH1a2 inhibitor

Inhibitor of aldehyde dehydrogenase 1a2 (ALDH1a2) (IC50 = 0.3 μM). Inhibits the biosynthesis of retinoic acid from retinol in neonatal and adult murine testis.

Chemical Identifiers

CAS 1477-57-2
Molecular Formula C12H20Cl4N2O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 366.11
Purity >98%
Molecular Weight (g/mol) 366.104
Chemical Name or Material WIN 18446
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.