Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ WIN 64338 hydrochloride
GREENER_CHOICESDP

Catalog No. 105750 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-575-0 50 mg
10-571-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 10-575-0 Supplier R&D Systems™ Supplier No. 1057/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Bradykinin B2 antagonist

The first potent, non-peptide, competitive bradykinin B2 receptor antagonist. In organ bath studies, WIN 64338 inhibits [3H]-bradykinin binding on guinea pig trachea with nanomolar affinity but is not active in the rabbit aorta (the classical bradykinin B1 preparation).

Chemical Identifiers

CAS 163727-74-0
Molecular Formula C45H69Cl2N4OP

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 783.95
Purity >98%
Molecular Weight (g/mol) 783.948
Chemical Name or Material WIN 64338 hydrochloride
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.