Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WIPF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570410
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570410 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570410 Supplier Invitrogen™ Supplier No. PA570410
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 79%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 93%.

This gene encodes a protein that plays an important role in the organization of the actin cytoskeleton. The encoded protein binds to a region of Wiskott-Aldrich syndrome protein that is frequently mutated in Wiskott-Aldrich syndrome, an X-linked recessive disorder. Impairment of the interaction between these two proteins may contribute to the disease. Two transcript variants encoding the same protein have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WIPF1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene WIPF1
Gene Accession No. Q6IN36
Gene Alias AI115543; D2Ertd120e; MGC111041; Protein PRPL-2; PRPL-2; testicular tissue protein Li 226; WAS/WASL interacting protein family member 1; WAS/WASL interacting protein family, member 1; WAS/WASL-interacting protein family member 1; WAS2; WASP interacting protein; WASP-interacting protein; WASPIP; Wip; Wipf1; Wiskott-Aldrich syndrome protein interacting protein; wiskott-Aldrich syndrome protein-interacting protein
Gene Symbols WIPF1
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of rat WIPF1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117538
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.