Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WIPF1/WIP Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | WIPF1/WIP |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
WIPF1/WIP Polyclonal specifically detects WIPF1/WIP in Rat samples. It is validated for Western Blot.Specifications
WIPF1/WIP | |
Western Blot | |
Unconjugated | |
Rabbit | |
Rat | |
WAS/WASL interacting protein family, member 1, WAS/WASL-interacting protein family member 1, WASP-interacting protein, WASPIP, WIPMGC111041, WiskProtein PRPL-2, WiskPRPL-2 | |
The immunogen is a synthetic peptide directed towards the N-terminal region of Rat WIPF1/WIP (NP_476540). Peptide sequence MPVPPPPAPPPPPTFALANTEKPSLNKTEQAGRNALLSDISKGKKLKKTV | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
7456 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title