Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WIPF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554948
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554948 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554948 Supplier Invitrogen™ Supplier No. PA554948
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MQRPSLPDLS RPNTTSSTGM KHSSSAPPPP PPGRRANAPP TPLPMHSSKA PAYNREKPLP PTPGQRLHPG REGPPAPP Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

This gene belongs to the GPI family whose members encode multifunctional phosphoglucose isomerase proteins involved in energy pathways. The protein encoded by this gene is a dimeric enzyme that catalyzes the reversible isomerization of glucose-6-phosphate and fructose-6-phosphate. The protein functions in different capacities inside and outside the cell. In the cytoplasm, the gene product is involved in glycolysis and gluconeogenesis, while outside the cell it functions as a neurotrophic factor for spinal and sensory neurons. Defects in this gene are the cause of nonspherocytic hemolytic anemia and a severe enzyme deficiency can be associated with hydrops fetalis, immediate neonatal death and neurological impairment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WIPF2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Wipf2
Gene Accession No. Q8TF74
Gene Alias 1110014J05Rik; 5730509C05Rik; AA407487; Gm1176; PP10631; RGD1561080; WAS/WASL interacting protein family member 2; WAS/WASL interacting protein family, member 2; WAS/WASL-interacting protein family member 2; WASP-binding protein; WASP-interacting protein-related protein; WICH; WIP- and CR16-homologous protein; WIPF2; WIP-related protein; Wire
Gene Symbols Wipf2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WIPF2. Recombinant protein control fragment (Product #RP-93449).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147179
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.