Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WIPF2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989411
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9411 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9411 Supplier Bioss Supplier No. BS9869R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene belongs to the GPI family whose members encode multifunctional phosphoglucose isomerase proteins involved in energy pathways. The protein encoded by this gene is a dimeric enzyme that catalyzes the reversible isomerization of glucose-6-phosphate and fructose-6-phosphate. The protein functions in different capacities inside and outside the cell. In the cytoplasm, the gene product is involved in glycolysis and gluconeogenesis, while outside the cell it functions as a neurotrophic factor for spinal and sensory neurons. Defects in this gene are the cause of nonspherocytic hemolytic anemia and a severe enzyme deficiency can be associated with hydrops fetalis, immediate neonatal death and neurological impairment.

Specifications

Antigen WIPF2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Wipf2
Gene Accession No. Q8TF74
Gene Alias 1110014J05Rik; 5730509C05Rik; AA407487; Gm1176; PP10631; RGD1561080; WAS/WASL interacting protein family member 2; WAS/WASL interacting protein family, member 2; WAS/WASL-interacting protein family member 2; WASP-binding protein; WASP-interacting protein-related protein; WICH; WIP- and CR16-homologous protein; WIPF2; WIP-related protein; Wire
Gene Symbols Wipf2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human WIPF2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147179
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.