Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WIPI1 Antibody (SR1849), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | WIPI1 |
---|---|
Clone | SR1849 |
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100 |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
WIPI1 Monoclonal antibody specifically detects WIPI1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
WIPI1 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Atg18, Atg18 protein homolog, ATG18A, FLJ10055, WD repeat domain phosphoinositide-interacting protein 1, WD repeat domain, phosphoinositide interacting 1, WD40 repeat protein interacting with phosphoinositides of 49 kDa, WD40 repeat protein Interacting with phosphoInositides of 49kDa, WIPI 49 kDa, WIPI-1, WIPI-1 alpha, WIPI49ATG18 | |
A synthesized peptide derived from human WIPI1 (Uniprot #: Q5MNZ9) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1849 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Autophagy, Cancer, Cell Biology, Endocrinology, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
55062 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title