Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WIPI1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110581
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110581 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110581 Supplier Invitrogen™ Supplier No. PA5110581
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GTLAAITFNA SGSKLASASE KGTVIRVFSV PDGQKLYEFR RGMKRYVTIS SLVFSMDSQF LCASSNTETV HIFKLEQVTN SRPEEPSTWS GYMGKMFMAA TNYLPTQVSD MMHQDRAFAT ARLNFSGQRN ICTLSTIQKL PRLLVASSSG HLYMYNLDPQ DGGECVLIKT HSLLGSGTTE ENKENDLRPS LPQSYAATVA RPSASSASTV PGYSEDGGAL RGEVIPEHEF ATGPVCLDDE NEFPPIILCR GNQKGKTKQS.

WIPI1 (WD repeat domain, phosphoinositide interacting-1), also known as WIPI1, ATG18 or WIPI49, is thought to play a role in autophagy and may regulate protein trafficking in certain recycling pathways. It contains three WD repeats and has a 7-bladed propeller structure with a conserved motif that facilitates its interaction with other proteins. WIPI1 localizes to cytoplasmic vesicles, endosomes, clathrin-coated vesicles and the trans-Golgi network. It is ubiquitously expressed with highest expression in heart, testis, placenta, pancreas and skeletal muscle. WIPI1 is upregulated in a variety of tumors, suggesting a role in carcinogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WIPI1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.46 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene WIPI1
Gene Accession No. Q5MNZ9, Q8R3E3
Gene Alias 4930533H01Rik; ATG18; Atg18 protein homolog; ATG18A; AW411817; D11Ertd498e; RGD1307754; WD repeat domain phosphoinositide-interacting protein 1; WD repeat domain, phosphoinositide interacting 1; WD40 repeat protein interacting with phosphoinositides of 49 kDa; WD40 repeat protein Interacting with phosphoInositides of 49kDa; WIPI 49 kDa; Wipi1; WIPI-1; WIPI-1 alpha; WIPI49
Gene Symbols WIPI1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 187-446 of human WIPI1 (NP_060453.3).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303630, 52639, 55062
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.