Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WIPI2 Antibody (2A2), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308559AF647
Description
WIPI2 Monoclonal antibody specifically detects WIPI2 in Human, Mouse samples. It is validated for Flow Cytometry, ELISASpecifications
WIPI2 | |
Monoclonal | |
Alexa Fluor 647 | |
ATG18B, Atg21, CGI-50, DKFZP434J154, DKFZp686P02188, FLJ12979, FLJ14217, FLJ42984, WD repeat domain phosphoinositide-interacting protein 2, WD repeat domain, phosphoinositide interacting 2, WD40 repeat protein interacting with phosphoinositides 2, WIPI-2, WIPI49-like protein 2 | |
Recombinant fragment (around aa 418-436) of human WIPI2 protein (exact sequence is proprietary) (Uniprot: Q9Y4P8) | |
100 μg | |
Autophagy, Cell Biology, Endocrinology | |
26100 | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry, ELISA | |
2A2 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction