Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WIPI2 Polyclonal Antibody

WIPI2 Polyclonal Antibody
Supplier: Thermo Scientific PA554425
Description
WIPI2 Polyclonal Antibody for Western Blot, ICC/IF, IHC

Specifications
WIPI2 | |
Polyclonal | |
Unconjugated | |
WIPI2 | |
1110018O08Rik; 2510001I10Rik; ATG18B; Atg21; CGI-50; WD repeat domain phosphoinositide-interacting protein 2; WD repeat domain, phosphoinositide interacting 2; WD40 repeat protein interacting with phosphoinositides 2; WIPI2; WIPI-2; WIPI49-like protein 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26100 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y4P8 | |
WIPI2 | |
Recombinant protein corresponding to Human WIPI2. Recombinant protein control fragment (Product #RP-92774). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction