Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WIPI2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554098
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554098 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554098 Supplier Invitrogen™ Supplier No. PA554098
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ECALMKQHRL DGSLETTNEI LDSASHDCPL VTQTYGAAAG KGTYVPSSPT RLAYTDDLGA VGGACLEDEA SALRLDEDSE HPPMILRTD Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 84%.

WD repeat proteins play a role in many essential biologic functions, regulating the assembly of multiprotein complexes by presenting a beta-propeller platform for simultaneous and reversible protein-protein interactions. WIPI2, also known as ATG18B or ATG21, is a human homolog to yeast ATG18 and contains three WD repeats and has a 7-bladed propeller structure with a conserved motif that facilitates its interaction with other proteins. It is recruited to early autophagosomal structures along with Atg16L and ULK1 and is required for the formation of LC3-positive autophagosomes. Along with the highly related WIPI1, WIPI2 is found at the plasma membrane in addition to autophagosomal membranes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WIPI2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WIPI2
Gene Accession No. Q6AY57, Q80W47, Q9Y4P8
Gene Alias 1110018O08Rik; 2510001I10Rik; ATG18B; Atg21; CGI-50; WD repeat domain phosphoinositide-interacting protein 2; WD repeat domain, phosphoinositide interacting 2; WD40 repeat protein interacting with phosphoinositides 2; WIPI2; WIPI-2; WIPI49-like protein 2
Gene Symbols WIPI2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WIPI2. Recombinant protein control fragment (Product #RP-92773).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26100, 288498, 74781
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.