Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Wire Gauge
SCI_ED

Catalog No. p-7075559 Shop All Eisco Products
Change view
Click to view available options
Length (Metric):
12.5 cm
12.7 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
S13971 12.5 cm
S13972 12.7 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S13971 Supplier Eisco™ Supplier No. CH0526A
Add to Cart
Add to Cart

For measuring the diameter of wire.

  • Made from iron
  • Plain

Specifications

Width (Metric) 127 mm
Edges Folded
Ceramic Center No Ceramic Center
Shape Square
For Use With (Application) Measuring the Diameter of Wire
Length (Metric) 12.5 cm
Material Iron
Type Wire Gauze
Description Plain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.