Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WISP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116124
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116124 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116124 Supplier Invitrogen™ Supplier No. PA5116124
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total WISP2.

Wisp2 a member of the WNT1 inducible signaling pathway (WISP) protein subfamily, which belongs to the connective tissue growth factor (CTGF) family. WNT1 is a member of a family of cysteine-rich, glycosylated signaling proteins that mediate diverse developmental processes. The CTGF family members are characterized by four conserved cysteine-rich domains: insulin-like growth factor-binding domain, von Willebrand factor type C module, thrombospondin domain and C-terminal cystine knot-like (CT) domain. Wisp2 lacks the CT domain which is implicated in dimerization and heparin binding. It is 72% identical to the mouse protein at the amino acid level. This gene may be downstream in the WNT1 signaling pathway that is relevant to malignant transformation. Its expression in colon tumors is reduced while the other two WISP members are overexpressed in colon tumors. It is expressed at high levels in bone tissue, and may play an important role in modulating bone turnover.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WISP2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene CCN5
Gene Accession No. O76076, Q9JHC6
Gene Alias CCN family member 5; CCN family protein COP-1; Ccn5; connective tissue growth factor-like protein; Connective tissue growth factor-related protein 58; Cop1; Crgr4; CT58; CTGFL; CTGF-L; Rcop1; UNQ228/PRO261; WISP2; WISP-2; WNT1 inducible signaling pathway protein 2; WNT1-inducible-signaling pathway protein 2
Gene Symbols CCN5
Host Species Rabbit
Immunogen A synthesized peptide derived from human WISP2(Accession O76076), corresponding to amino acid residues T214-F250.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29576, 8839
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.