Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WIZ Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP121059
Description
WIZ Polyclonal specifically detects WIZ in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Peptide ELISA.Specifications
WIZ | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
widely interspaced zinc finger motifs, widely-interspaced zinc finger motifs, Widely-interspaced zinc finger-containing protein, WIZ zinc finger, Zinc finger protein 803, ZNF803protein Wiz | |
Goat | |
85 kDa | |
0.1 mg | |
Primary | |
Expected from sequence similarity: Rat. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:2000 | |
O95785 | |
WIZ | |
Peptide with sequence C-RSYIQGGRPFTKKFR corresponding to internal region according to NP_067064.2. | |
Affinity Purified | |
RUO | |
58525 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction