Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WIZ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582844
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582844 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582844 Supplier Invitrogen™ Supplier No. PA582844
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LKKKSKPCLI KKEPPAGDLA PALAEDGPPT VAPGPVQSPL PLSPLAGRPG KPGAGPAQVP RELSLTPITG AKPSATGYLG SVAAKRPLQE DRLLPAEVKA KTYIQTELPF KAKTLHEKTS HSS Highest antigen sequence indentity to the following orthologs: Mouse - 87%, Rat - 88%.

WIZ, also known as ZNF803, was initially identified in a mouse cerebellum cDNA library screen and its message was found to be expressed in the granule cell layers of the cerebellum as well as in the dentate gyrus and olfactory bulb. Later analysis indicates however that WIZ is ubiquitously expressed. WIZ is a nuclear protein that co-localizes with G9a, a histone methyltransferase responsible for mono- and dimethylation of H3K9 at euchromatic regions. WIZ can also associate with CtBP family proteins, known to be transcriptional co-repressors, and has been suggested to link G9a/GLP complexes to the CtBP co-repressor machinery, possibly regulating complex stability and gene silencing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WIZ
Applications ChIP Assay, Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WIZ
Gene Accession No. O95785
Gene Alias Protein Wiz; widely interspaced zinc finger motifs; widely-interspaced zinc finger motifs; widely-interspaced zinc finger-containing protein; Wiz; WIZ zinc finger; zinc finger protein 803; ZNF803
Gene Symbols WIZ
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WIZ. Recombinant protein control fragment (Product #RP-104572).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58525
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.