Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Wizard™ Magnetic 96 DNA Plant System

Catalog No. PRA3811 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Product Type:
96 DNA Plant System
Replacement Wash Buffer
Quantity:
4 x 96 Preps
40 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
PRA3811 Replacement Wash Buffer 40 mL
PRFF3761 96 DNA Plant System 4 x 96 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRA3811 Supplier Promega Supplier No. A3811
Add to Cart
Add to Cart

Provides manual or automated 96-well, high-throughput purification of DNA from plant leaf and seed tissue.

  • Provides manual or automated 96-well, high-throughput purification of DNA from plant leaf and seed tissue
  • More rapid than CTAB extraction
  • Eliminates organic extractions, multiple centrifugations, and cumbersome filter plates
  • Allows nucleic acids to bind in solution, increasing purity of final product
  • Verified for both leaf and seed tissue by PCR and RAPD analysis
  • Compatible with many robotic platforms

Specifications

Content And Storage 15°C to 30°C
Product Type Replacement Wash Buffer
Quantity 40 mL
Sample Volume 40mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.