Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Wizard™ Magnetic 96 DNA Plant System

Catalog No. p-3081962
Click to view available options
Product Type:
96 DNA Plant System
Replacement Wash Buffer
Quantity:
4 x 96 Preps
40 mL

Provides manual or automated 96-well, high-throughput purification of DNA from plant leaf and seed tissue.

  • Automated Purification of DNA from Leaves and Seeds
  • Cuts purification time compared to CTAB extraction
  • Eliminates organic extractions.
  • Validated for both leaf and seed tissue by PCR and RAPD analysis

Specifications

Content And Storage 15°C to 30°C
No. of Reactions 4 x 96 Preps
Product Type 96 DNA Plant System
Quantity 4 x 96 Preps
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.