Learn More
Invitrogen™ WNK1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120255
Description
Positive test controls include: Mouse liver, Rat liver. The target is usually found in the following locations: Cytoplasm. Immunogen sequence: MSGGAAEKQS STPGSLFLSP PAPAPKNGSS SDSSVGEKLG AAAADAVTGR TEEYRRRRHT MDKDSRGAAA TTTTTEHRFF RRSVICDSNA TALELPGLPL SLPQPSIPAA VPQSAPPEPH REETVTATAT SQVAQQPPAA AAPGEQAVAG PAPSTVPSST SKDRPVSQPS LVGSKEEPPP ARSGSGGGSA KEPQEERSQQ QDDIEELETK.
WNK1 serine/threonine kinase is a member of the WNK subfamily. WNK1 may be a key regulator of blood pressure by controlling the transport of sodium and chloride ions. Mutations in this gene have been associated with pseudohypoaldosteronism type II and hereditary sensory neuropathy type II. WNK plays an important role in the regulation of electrolyte homeostasis, cell signaling, survival, and proliferation. Also, acts as an activator and inhibitor of sodium-coupled chloride cotransporters and potassium-coupled chloride cotransporters respectively. Activates SCNN1A, SCNN1B, SCNN1D, and SGK1. WNK localizes to a segment of kidney involved in salt, potassium, and pH homeostasis, the distal nephron. Mutations in the WNK gene result in Pseudohypaldosteronism and Neuropathy.
Specifications
| WNK1 | |
| Polyclonal | |
| Unconjugated | |
| WNK1 | |
| 6430573H23Rik; EG406236; Erythrocyte 65 kDa protein; HSAN2; HSN2; hWNK1; KDP; KIAA0344; Kinase deficient protein; MGC163339; MGC163341; mKIAA0344; p65; PPP1R167; Prkwnk1; prostate-derived sterile 20-like kinase; protein kinase lysine-deficient 1; Protein kinase with no lysine 1; protein phosphatase 1, regulatory subunit 167; PSK; Serine/threonine-protein kinase WNK1; serine/threonine-protein kinase WNK1 1; serine/threonine-protein kinase WNK1 2; With no K; WNK lysine deficient protein kinase 1; WNK lysine deficient protein kinase 1 isoform; WNK1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 116477, 232341 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.44 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P83741, Q9JIH7 | |
| WNK1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human WNK1 (NP_055638.2). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.