Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WNK3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596362
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596362 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596362 Supplier Invitrogen™ Supplier No. PA596362
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Rat hypothalamus, Rat CNS, Rat brain; Cellular Location: Cytoplasm.

WNK3 is a serine/threonine kinase involved in signal transduction and cellular communication. Members of the 'with no lysine' kinase family, such as WNK3, are serine-threonine protein kinases that lack the almost invariant catalytic lysine in subdomain II, which is important for binding ATP in the catalytic site. Instead, these kinases have a conserved lysine in subdomain I that is thought to provide this function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WNK3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.98 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene WNK3
Gene Accession No. Q80XP9, Q9BYP7
Gene Alias KIAA1566; PRKWNK 3; PRKWNK3; Protein kinase lysine-deficient 3; protein kinase with no lysine 3; RGD1563131; serine/threonine-protein kinase WNK3; WNK 3; WNK lysine deficient protein kinase 3; WNK lysine deficient protein kinase 3, pseudogene; WNK3; Wnk3-ps
Gene Symbols WNK3
Host Species Rabbit
Immunogen Recombinant protein of rat WNK3.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 279561, 317420, 65267
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.