Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WNK3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567465
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567465 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567465 Supplier Invitrogen™ Supplier No. PA567465
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSQPSVAYSSN QTMGSQMVSN IPQAEVNVPG QIYSSQQLVG HYQQVSGLQK HSKLTQPQIL PLVQGQSTVL PVHVLGPTVV SQPQVSPLTV QKV Highest antigen sequence identity to the following orthologs - mouse 59%, rat 41%.

WNK3 is a serine/threonine kinase involved in signal transduction and cellular communication. Members of the 'with no lysine' kinase family, such as WNK3, are serine-threonine protein kinases that lack the almost invariant catalytic lysine in subdomain II, which is important for binding ATP in the catalytic site. Instead, these kinases have a conserved lysine in subdomain I that is thought to provide this function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WNK3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WNK3
Gene Accession No. Q9BYP7
Gene Alias KIAA1566; PRKWNK 3; PRKWNK3; Protein kinase lysine-deficient 3; protein kinase with no lysine 3; RGD1563131; serine/threonine-protein kinase WNK3; WNK 3; WNK lysine deficient protein kinase 3; WNK lysine deficient protein kinase 3, pseudogene; WNK3; Wnk3-ps
Gene Symbols WNK3
Host Species Rabbit
Immunogen Recombinant Human WNK3. Recombinant protein control fragment (Product #RP-108120).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65267
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.