Learn More
WNK3 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301877A
Description
The recommended shelf life for this product is 1 year from date of receipt.
Members of the ′with no lysine′ kinase family, such as WNK3, are serine-threonine protein kinases that lack the almost invariant catalytic lysine in subdomain II, which is important for binding ATP in the catalytic site. Instead, these kinases have a conserved lysine in subdomain I that is thought to provide this function.Specifications
| WNK3 | |
| Polyclonal | |
| Unconjugated | |
| WNK3 | |
| WNK lysine deficient protein kinase 3, KIAA1566, PRKWNK3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 65267 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q9BYP7 | |
| WNK3 | |
| Between 1 and 50 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.