Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WNK4 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB600284APC
Description
WNK4 Polyclonal antibody specifically detects WNK4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunomicroscopy, ICC/IF (Negative)Specifications
WNK4 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Electron Microscopy, ICC/IF (Negative) | |
EC 2.7.11, EC 2.7.11.1, PHA2B, PRKWNK4protein kinase, lysine deficient 4, Protein kinase lysine-deficient 4, Protein kinase with no lysine 4, serine/threonine-protein kinase WNK4, WNK lysine deficient protein kinase 4 | |
A synthetic peptide made to a region of the human WNK4 protein between residues 200-300. [UniProt# Q96J92] | |
0.1 mL | |
Cell Biology, Potassium Channels, Protein Kinase, Signal Transduction | |
65266 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Immunomicroscopy | |
APC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction