Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WNK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. APPA6878
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
APPA6878 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. APPA6878 Supplier Invitrogen™ Supplier No. PA116878
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is not recommended for ICC/IF applications. Suggested positive control: mouse kidney lysate or protein.

This gene encodes a member of the WNK family of serine-threonine protein kinases. The kinase is part of the tight junction complex in kidney cells, and regulates the balance between NaCl reabsorption and K(+) secretion. The kinase regulates the activities of several types of ion channels, cotransporters, and exchangers involved in electrolyte flux in epithelial cells. Mutations in this gene result in pseudohypoaldosteronism type IIB.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WNK4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Wnk4
Gene Accession No. Q7TPK6, Q80UE6, Q96J92
Gene Alias 2010002J11Rik; Ac2-059; Pha2b; Prkwnk4; protein kinase lysine-deficient 4; Protein kinase with no lysine 4; protein kinase, lysine deficient 4; protein kinase, lysine-deficient 4; RP23-281C18.8; serine/threonine-protein kinase WNK4; WNK lysine deficient protein kinase 4; WNK4; WNK4 Ser/Thr kinase
Gene Symbols Wnk4
Host Species Rabbit
Immunogen A synthetic peptide made to a region of the human WNK4 protein between residues 200-300.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 287715, 65266, 69847
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.