Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wnt-1 Antibody (13F9), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP142739AF405
Description
Wnt-1 Monoclonal specifically detects Wnt-1 in Human, Mouse samples. It is validated for Western Blot, ELISA.Specifications
Wnt-1 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, ELISA | |
13F9 | |
Western Blot, ELISA | |
INT1Proto-oncogene Int-1 homolog, proto-oncogene Wnt-1, wingless-type MMTV integration site family, member 1, wingless-type MMTV integration site family, member 1 (oncogene INT1) | |
Synthetic peptide corresponding to an internal region of human Wnt-1 protein. (Uniprot: P04628) | |
0.1 mL | |
Apoptosis, Breast Cancer, Cellular Markers, Oncogenes, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
7471 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction