Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wnt-1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310925100UL
Description
Wnt-1 Polyclonal specifically detects Wnt-1 in Mouse samples. It is validated for Western Blot.Specifications
| Wnt-1 | |
| Polyclonal | |
| Western Blot 1.0 ug/ml | |
| INT1Proto-oncogene Int-1 homolog, proto-oncogene Wnt-1, wingless-type MMTV integration site family, member 1, wingless-type MMTV integration site family, member 1 (oncogene INT1) | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of MOUSE Wnt-1 (NP_067254). Peptide sequence NFCTYSGRLGTAGTAGRACNSSSPALDGCELLCCGRGHRTRTQRVTERCN | |
| 100 μg | |
| Apoptosis, Breast Cancer, Cellular Markers, Oncogenes, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
| 7471 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| PBS buffer, 2% sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction