Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wnt-2 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306123UV
Description
Wnt-2 Polyclonal specifically detects Wnt-2 in Human samples. It is validated for Western Blot.Specifications
Wnt-2 | |
Polyclonal | |
Western Blot | |
INT1L1secreted growth factor, Int-1-like protein 1, Int-1-related protein, IRPprotein Wnt-2, wingless-type MMTV integration site family member 2 | |
Produced in rabbits immunized with E. coli-derived Human Wnt-2 fragment. | |
0.1 mL | |
Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
7472 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction