Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wnt-2 Antibody (HL1999) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP325752
Description
Wnt-2 Monoclonal antibody specifically detects Wnt-2 in Human, Mouse, Rat, Zebrafish samples. It is validated for Western BlotSpecifications
Wnt-2 | |
Monoclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Zebrafish | |
Purified |
Western Blot | |
HL1999 | |
Western Blot 1:5000-1:20000 | |
INT1L1secreted growth factor, Int-1-like protein 1, Int-1-related protein, IRPprotein Wnt-2, wingless-type MMTV integration site family member 2 | |
Recombinant fragment of human Wnt-2. | |
100 μL | |
Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
7472 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction