Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wnt-2 Antibody (SR1571), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Wnt-2 |
---|---|
Clone | SR1571 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
Wnt-2 Monoclonal antibody specifically detects Wnt-2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Wnt-2 | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
INT1L1secreted growth factor, Int-1-like protein 1, Int-1-related protein, IRPprotein Wnt-2, wingless-type MMTV integration site family member 2 | |
A synthesized peptide derived from human Wnt-2 (Uniprot #: P09544) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1571 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
7472 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title