Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wnt-3a Antibody (217804.2R) [Alexa Fluor™ 532], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB13242X
Description
Wnt-3a Monoclonal antibody specifically detects Wnt-3a in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOFSpecifications
Wnt-3a | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
217804.2R | |
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
MGC119418, MGC119419, MGC119420, protein Wnt-3a, wingless-type MMTV integration site family, member 3A | |
E.coli-derived recombinant mouse Wnt-3a, Ser36-Gln75, Trp219-Arg269, Accession # P27467 | |
0.1 mL | |
Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
89780 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction