Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wnt-5a Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP224752UV
Description
Wnt-5a Polyclonal specifically detects Wnt-5a in Human, Mouse, Rat, Bovine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Wnt-5a | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
hWNT5A, protein Wnt-5a, wingless-type MMTV integration site family, member 5A, WNT-5A protein | |
Rabbit | |
Affinity Purified | |
Cancer Stem Cells, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
7474 | |
Store at 4C in the dark. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
WNT5A | |
A synthetic peptide corresponding to amino acids 190-230 of human Wnt-5a was used as the immunogen for this antibody, GenBank no NP_003383.2. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Bovine, Monkey | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction