Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wnt-5b Antibody (SR1784), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Wnt-5b |
---|---|
Clone | SR1784 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50 |
Applications | Western Blot, Flow Cytometry |
Classification | Monoclonal |
Description
Wnt-5b Monoclonal antibody specifically detects Wnt-5b in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
Wnt-5b | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
MGC2648, protein Wnt-5b, wingless-type MMTV integration site family, member 5B, WNT-5B protein | |
A synthesized peptide derived from human Wnt-5b (Uniprot #: Q9H1J7) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1784 | |
Western Blot, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
81029 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title