Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wnt-9a Antibody (326711), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB3157X
Description
Wnt-9a Monoclonal antibody specifically detects Wnt-9a in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOFSpecifications
Wnt-9a | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
326711 | |
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
MGC138165, MGC141991, Protein Wnt-14, wingless-type MMTV integration site family, member 14, wingless-type MMTV integration site family, member 9A, WNT14protein Wnt-9a | |
E. coli-derived recombinant human Wnt-9a, Pro48-Glu92&Arg231-Phe304, Accession # O14904.2 | |
0.1 mL | |
Signal Transduction, Wnt Signaling Pathway | |
7483 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction