Learn More
WNT1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | WNT1 |
---|---|
Concentration | 300 μg/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-8596
![]() ![]() |
Proteintech Group Inc
279351AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The WNT1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human WNT1. This antibody recognizes human, mouse antigen. The WNT1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
WNT1 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Human, Mouse | |
IgG |
300 μg/mL | |
Polyclonal | |
Rabbit | |
7471 | |
Primary |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.